Thermo Scientific Chemicals

Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals

Catalog number: 420710250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals

Catalog number: 420710250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 420710250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5810-42-4
IUPAC Name
tetrapropylazanium chloride
Molecular Formula
C12H28ClN
InChI Key
FBEVECUEMUUFKM-UHFFFAOYSA-M
SMILES
[Cl-].CCC[N+](CCC)(CCC)CCC
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Titration Argentometric
>=92 %
Appearance (Form)
Crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text