Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals
Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 420711000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametetrapropylazanium chloride
Molecular FormulaC12H28ClN
InChI KeyFBEVECUEMUUFKM-UHFFFAOYSA-M
SMILES[Cl-].CCC[N+](CCC)(CCC)CCC
Molecular Weight (g/mol)221.81
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration Argentometric>=92 %
Appearance (Form)Crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only