Thermo Scientific Chemicals

Tetrapropylammonium hydroxide, 25% in water, Thermo Scientific Chemicals

Catalog number: 420730250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tetrapropylammonium hydroxide, 25% in water, Thermo Scientific Chemicals

Catalog number: 420730250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 420730250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4499-86-9
IUPAC Name
tetrapropylazanium hydroxide
Molecular Formula
C12H29NO
InChI Key
LPSKDVINWQNWFE-UHFFFAOYSA-M
SMILES
[OH-].CCC[N+](CCC)(CCC)CCC
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
Titration with HClO4
24 to 26 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text