Thermo Scientific Chemicals

Thallic trifluoroacetate, tech., Thermo Scientific Chemicals

Catalog number: 420760250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Thallic trifluoroacetate, tech., Thermo Scientific Chemicals

Catalog number: 420760250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420760250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23586-53-0
IUPAC Name
thallium(3+) tritrifluoroacetate
Molecular Formula
C6F9O6Tl
InChI Key
PSHNNUKOUQCMSG-UHFFFAOYSA-K
SMILES
[Tl+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Appearance (Form)
Powder
Appearance (Color)
Light yellow to beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text