Thermo Scientific Chemicals

2-Thiophenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 420890250
25 g, Each
Thermo Scientific Chemicals

2-Thiophenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 420890250
25 g, Each
Quantity
Catalog number: 420890250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16629-19-9
IUPAC Name
thiophene-2-sulfonyl chloride
Molecular Formula
C4H3ClO2S2
InChI Key
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=CS1
Infrared spectrum
Conforms
Appearance (Color)
Colorless to blue-green to brown-purple
Titration Argentometric
>=97.5 % (Ex Cl)
Appearance (Form)
Low melting solid or liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text