Thermo Scientific Chemicals

3,6-Thioxanthenediamine-10,10-dioxide, 97%, Thermo Scientific Chemicals

Catalog number: 420910100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

3,6-Thioxanthenediamine-10,10-dioxide, 97%, Thermo Scientific Chemicals

Catalog number: 420910100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420910100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10215-25-5
IUPAC Name
3,6-diamino-9H-10λ⁶-thioxanthene-10,10-dione
Molecular Formula
C13H12N2O2S
InChI Key
UPVRZVIJGVFROW-UHFFFAOYSA-N
SMILES
NC1=CC=C2CC3=CC=C(N)C=C3S(=O)(=O)C2=C1
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
214°C to 219°C
Appearance (Color)
Beige to yellow-green
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text