Thermo Scientific Chemicals

Thymol Blue Sodium Salt, For analysis ACS

Catalog number: 420940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Thymol Blue Sodium Salt, For analysis ACS

Catalog number: 420940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420940250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62625-21-2
IUPAC Name
sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
Molecular Formula
C27H29NaO5S
InChI Key
OCMIKNBSIRSUPI-FRWNMSGJSA-M
SMILES
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
Clarity of solution
Passes test
Infrared spectrum
Conforms
Visual transition interval
From pH 1.2 (pink) to pH 2.8 (yellow)
Appearance (Form)
Powder
Visual transition interval
From pH 8.0 (yellow) to pH 9.2 (blue)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text