Chemical Identifiers
CAS125-20-2
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H30O4
InChI KeyLDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more