Thermo Scientific Chemicals

Tin, 30 mesh, Reagent ACS, Granular, Thermo Scientific Chemicals

Catalog number: 420995000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tin, 30 mesh, Reagent ACS, Granular, Thermo Scientific Chemicals

Catalog number: 420995000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420995000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-31-5
IUPAC Name
tin
Molecular Formula
Sn
InChI Key
ATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES
[Sn]
Appearance (Color)
Grey
Appearance (Form)
Metallic granular powder
Titration Complexometric
>=99.5 %
Antimony (Sb)
=<200 ppm
Arsenic (As)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text