1x1 image pixel for data collection
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
Thermo Scientific Chemicals

D-gamma-Tocopherol, 95%

CAS: 54-28-4 | C28H48O2 | 416.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
421041000100 mgGlass bottle
Catalog number 421041000
Price (USD)/ Each
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Add to cart
Chemical Identifiers
IUPAC Name(2R)-2,7,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular FormulaC28H48O2
InChI KeyQUEDXNHFTDJVIY-DQCZWYHMSA-N
SMILESCC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=C(C)C(C)=C2O1
Molecular Weight (g/mol)416.69
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to light brown
Appearance (Form)Viscous liquid
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link