Thermo Scientific Chemicals

D-δ-Tocopherol, 93%, Thermo Scientific Chemicals

Catalog number: 421055000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

D-δ-Tocopherol, 93%, Thermo Scientific Chemicals

Catalog number: 421055000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 421055000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
119-13-1
IUPAC Name
(2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular Formula
C27H46O2
InChI Key
GZIFEOYASATJEH-VHFRWLAGSA-N
SMILES
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1
Appearance (Color)
Clear yellow to brown
Appearance (Form)
Viscous liquid
GC
>=91 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text