DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
Thermo Scientific Chemicals

DL-alpha-Tocopheryl acetate, 98%

CAS: 7695-91-2 | C31H52O3 | 472.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 421070050
Price (USD)
150.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
150.00
Each
Add to cart
Chemical Identifiers
CAS7695-91-2
IUPAC Name(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Molecular FormulaC31H52O3
InChI KeyZAKOWWREFLAJOT-CEFNRUSXSA-N
SMILESCC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear light yellow to light green
GC>=97.5 %
Appearance (Form)Viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0457530Certificate of AnalysisJan 30, 2025421070010, 421070050
A0457530Certificate of AnalysisNov 16, 2023421070010, 421070050
A0420546Certificate of AnalysisAug 25, 2023421070010, 421070050
A0404356Certificate of AnalysisDec 13, 2022421070010, 421070050
A0409874Certificate of AnalysisDec 13, 2022421070010, 421070050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection