p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number 421210250
Price (USD)/ Each
24.65
Online exclusive
27.30 
Save 2.65 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
24.65
Online exclusive
27.30 
Save 2.65 (10%)
Add to cart
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Catalog number421210250
Price (USD)/ Each
24.65
Online exclusive
27.30 
Save 2.65 (10%)
-
Add to cart
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Clarity of solution(c=20, water) Passes test
Heavy metals (as Pb)=<0.001 %
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only