Thermo Scientific Chemicals

p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals

Catalog number: 421231000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

p-Toluenesulfonic acid, sodium salt, Thermo Scientific Chemicals

Catalog number: 421231000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 421231000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
657-84-1
IUPAC Name
sodium 4-methylbenzene-1-sulfonate
Molecular Formula
C7H7NaO3S
InChI Key
KVCGISUBCHHTDD-UHFFFAOYSA-M
SMILES
[Na+].CC1=CC=C(C=C1)S([O-])(=O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Titration after Ion exchange
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text