Thermo Scientific Chemicals

Tributyl citrate, 99+%, Thermo Scientific Chemicals

Catalog number: 421440010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Tributyl citrate, 99+%, Thermo Scientific Chemicals

Catalog number: 421440010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 421440010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-94-1
IUPAC Name
1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C18H32O7
InChI Key
ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
SMILES
CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
Appearance (Form)
Clear liquid
Refractive index
1.4440 to 1.4460 (20°C, 589 nm)
Color scale
=<50 APHA
GC
>=99.0 %
Acidity
=<0.2 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text