Thermo Scientific Chemicals

Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 421450025
2.5 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Trichloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 421450025
2.5 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 421450025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-03-9
IUPAC Name
trichloroacetic acid
Molecular Formula
C2HCl3O2
InChI Key
YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES
OC(=O)C(Cl)(Cl)Cl
Appearance (Color)
White
Heavy metals (as Pb)
=<20 ppm
Nitrate (NO3)
=<20 ppm
Phosphate (PO4)
=<5 ppm
Clarity of solution
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text