Thermo Scientific Chemicals

Trichloroethylene, 99.6%, ACS reagent, stabilized, Thermo Scientific Chemicals

Catalog number: 421520025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Trichloroethylene, 99.6%, ACS reagent, stabilized, Thermo Scientific Chemicals

Catalog number: 421520025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 421520025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-01-6
IUPAC Name
1,1,2-trichloroethene
Molecular Formula
C2HCl3
InChI Key
XSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILES
ClC=C(Cl)Cl
Color scale
=<10 APHA
Water
=<0.02 % (Coulometric)
Titrable acid
=<0.0001 meq/g
Free halogens
Passes test
Heavy metals (as Pb)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text