Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Glass Bottle
Catalog number 421540010
Price (USD)/ Each
79.65
Online exclusive
93.50 
Save 13.85 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
93.50 
Save 13.85 (15%)
Add to cart
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Catalog number421540010
Price (USD)/ Each
79.65
Online exclusive
93.50 
Save 13.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrichloro-1,3,5-triazinane-2,4,6-trione
Molecular FormulaC3Cl3N3O3
InChI KeyYRIZYWQGELRKNT-UHFFFAOYSA-N
SMILESClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
Molecular Weight (g/mol)232.40
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration Iodometric>=98.5 %
Appearance (Form)Powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only