Thermo Scientific Chemicals

Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 421610050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tricyclohexylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 421610050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 421610050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2622-14-2
IUPAC Name
tricyclohexylphosphane
Molecular Formula
C18H33P
InChI Key
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILES
C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Appearance (Color)
White to light yellow
NMR (31P-NMR)
>=96.0 %
Appearance (Form)
Crystalline powder
Melting point
77°C to 83°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text