Thermo Scientific Chemicals

Tridodecylamine, 90%, Thermo Scientific Chemicals

Catalog number: 421620250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tridodecylamine, 90%, Thermo Scientific Chemicals

Catalog number: 421620250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 421620250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
102-87-4
IUPAC Name
tridodecylamine
Molecular Formula
C36H75N
InChI Key
SWZDQOUHBYYPJD-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4550 to 1.4610 (20°C, 589 nm)
Infrared spectrum
Conforms
Titration with HClO4
>=88 %
Appearance (Form)
Liquid or low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text