Thermo Scientific Chemicals

Trimethylamine N-oxide dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 421891000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Trimethylamine N-oxide dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 421891000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 421891000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62637-93-8
IUPAC Name
N,N-dimethylmethanamine oxide dihydrate
Molecular Formula
C3H13NO3
InChI Key
PGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILES
O.O.C[N+](C)(C)[O-]
Melting point
94°C to 98°C
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystalline powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text