Trimethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals
Trimethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number 421960100
Price (USD)/ Each
68.65
Online exclusive
76.60 
Save 7.95 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
76.60 
Save 7.95 (10%)
Add to cart
Trimethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals
Catalog number421960100
Price (USD)/ Each
68.65
Online exclusive
76.60 
Save 7.95 (10%)
-
Add to cart
Chemical Identifiers
CAS420-37-1
IUPAC Nametetrafluoroboranuide; trimethyloxidanium
Molecular FormulaC3H9BF4O
InChI KeyCZVZBKHWOFJNCR-UHFFFAOYSA-N
SMILESC[O+](C)C.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Titration with NaOH94.0 to 106.0 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only