2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 421980025
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
Add to cart
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Catalog number421980025
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
-
Add to cart
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Appearance (Form)Clear liquid
UVat 210 nm A: =<1.0
UVat 220 nm A: =<0.20
UVat 230 nm A: =<0.10
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only