Trimyristin, 90%, Thermo Scientific Chemicals
Trimyristin, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimyristin, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 422090250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS555-45-3
IUPAC Name1,3-bis(tetradecanoyloxy)propan-2-yl tetradecanoate
Molecular FormulaC45H86O6
InChI KeyDUXYWXYOBMKGIN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
TLC>=88 %
Appearance (Form)Crystalline powder
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only