Thermo Scientific Chemicals

Tris(4-methoxyphenyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 422240100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(4-methoxyphenyl)phosphine, 95%, Thermo Scientific Chemicals

Catalog number: 422240100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 422240100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
855-38-9
IUPAC Name
tris(4-methoxyphenyl)phosphane
Molecular Formula
C21H21O3P
InChI Key
UYUUAUOYLFIRJG-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)P(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
Appearance (Form)
Crystalline powder
Melting point
131°C to 134°C
Infrared spectrum
Conforms
GC
>=94 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text