Thermo Scientific Chemicals

Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals

Catalog number: 422255000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals

Catalog number: 422255000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 422255000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14634-91-4
IUPAC Name
λ²-iron(2+) tris(1,10-phenanthroline) sulfate
Molecular Formula
C36H24FeN6O4S
InChI Key
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
SMILES
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Appearance (Color)
Dark red
Concentration
0.024 to 0.026 M (w/v in water)
Suitability
(for use as ind. for Fe) Passes test
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text