Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals
Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 422320050
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
Add to cart
Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals
Catalog number422320050
Price (USD)/ Each
85.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
100.00 
Save 14.35 (14%)
-
Add to cart
Chemical Identifiers
CAS6163-58-2
IUPAC Nametris(2-methylphenyl)phosphane
Molecular FormulaC21H21P
InChI KeyCOIOYMYWGDAQPM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point122°C to 126°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only