Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals
Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-o-tolylphosphine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 422320250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametris(2-methylphenyl)phosphane
Molecular FormulaC21H21P
InChI KeyCOIOYMYWGDAQPM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
Molecular Weight (g/mol)304.37
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point122°C to 126°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only