Tri-p-tolylphosphine, 98%, Thermo Scientific Chemicals
Tri-p-tolylphosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-p-tolylphosphine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 422330010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametris(4-methylphenyl)phosphane
Molecular FormulaC21H21P
InChI KeyWXAZIUYTQHYBFW-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Molecular Weight (g/mol)304.37
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point144°C to 148°C
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only