Thermo Scientific Chemicals

Triton™ X-100, scintillation grade

Catalog number: 422355000
500 mL, EcoSafPak™, Each
Thermo Scientific Chemicals

Triton™ X-100, scintillation grade

Catalog number: 422355000
500 mL, EcoSafPak™, Each
Quantity
Packaging
Catalog number: 422355000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-93-1
IUPAC Name
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI Key
JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonym
triton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
Refractive index
1.4900 to 1.4920 (20°C, 589 nm)
Infrared spectrum
Conforms
Color scale
=<100 APHA
Cloud point
63°C to 69°C (1% aq. soln.)
Appearance (Form)
Clear to slightly hazy liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text