Thermo Scientific Chemicals

Valeramide, 97%, Thermo Scientific Chemicals

Catalog number: 422440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Valeramide, 97%, Thermo Scientific Chemicals

Catalog number: 422440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 422440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
626-97-1
IUPAC Name
pentanamide
Molecular Formula
C5H11NO
InChI Key
IPWFJLQDVFKJDU-UHFFFAOYSA-N
SMILES
CCCCC(N)=O
Melting point
101°C to 106°C
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Glistening powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text