Thermo Scientific Chemicals

Vanadyl phthalocyanine, 85%, pract., Thermo Scientific Chemicals

Catalog number: 422491000
100 g, Each
Thermo Scientific Chemicals

Vanadyl phthalocyanine, 85%, pract., Thermo Scientific Chemicals

Catalog number: 422491000
100 g, Each
Quantity
Catalog number: 422491000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13930-88-6
IUPAC Name
2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide; oxovanadiumbis(ylium)
Molecular Formula
C32H16N8OV
InChI Key
YRZZLAGRKZIJJI-UHFFFAOYSA-N
SMILES
O=[V++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
Appearance (Form)
Powder
Infrared spectrum
Conforms
Appearance (Color)
Dark blue to purple
Assay
>=80 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text