Thermo Scientific Chemicals

4-Vinylbenzyl acetate, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 422500050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Vinylbenzyl acetate, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 422500050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 422500050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1592-12-7
IUPAC Name
(4-ethenylphenyl)methyl acetate
Molecular Formula
C11H12O2
InChI Key
LEIKPUSDAWATBV-UHFFFAOYSA-N
SMILES
CC(=O)OCC1=CC=C(C=C)C=C1
Appearance (Form)
Liquid
Refractive index
1.5355 to 1.5385 (20°C, 589 nm)
Stabilizer
0.1 % hydroquinone
Appearance (Color)
Clear light yellow to light pink
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text