Thermo Scientific Chemicals

Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals

Catalog number: 422680250
25 L, Each
Thermo Scientific Chemicals

Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals

Catalog number: 422680250
25 L, Each
Quantity
Catalog number: 422680250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1330-20-7
IUPAC Name
1,3-xylene
Molecular Formula
C8H10
InChI Key
IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES
CC1=CC(C)=CC=C1
GC
>=98.5 %
GC
Xylene isomers plus Ethylbenzene (Ethylbenzene
GC
not to exceed 25%)
Water
=<0.05 % (Coulometric)
Refractive index
1.4960 to 1.4980 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text