Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals
Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
4 L
20 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 422685000
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.20 
Save 7.55 (14%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.20 
Save 7.55 (14%)
Add to cart
Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals
Catalog number422685000
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.20 
Save 7.55 (14%)
-
Add to cart
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
GCXylene isomers plus Ethylbenzene (Ethylbenzene
GCnot to exceed 25%)
Water=<0.05 % (Coulometric)
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only