Thermo Scientific Chemicals

Xylene Cyanol FF 85%, Thermo Scientific Chemicals

Catalog number: 422690500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Xylene Cyanol FF 85%, Thermo Scientific Chemicals

Catalog number: 422690500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 422690500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4463-44-9
IUPAC Name
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Molecular Formula
C25H29N2NaO7S2
InChI Key
HLEQELPTFNOLFE-ZCTWALFCSA-N
SMILES
[H-].[Na+].CCNC1=CC=C(C=C1C)C(=C1/C=CC(=NCC)C(C)=C1)\C1=CC(O)=C(C=C1S(O)(=O)=O)S(O)(=O)=O
Appearance (Color)
Dark red to dark blue to purple
Appearance (Form)
Powder
Infrared spectrum
Conforms
Dye content
>=80 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text