Zinc oxide, ACS reagent, Thermo Scientific Chemicals
Zinc oxide, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc oxide, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 422741000
Price (USD)/ Each
20.65
Online exclusive
23.20 
Save 2.55 (11%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
20.65
Online exclusive
23.20 
Save 2.55 (11%)
Add to cart
Zinc oxide, ACS reagent, Thermo Scientific Chemicals
Catalog number422741000
Price (USD)/ Each
20.65
Online exclusive
23.20 
Save 2.55 (11%)
-
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
Titration Complexometric>=99.0 %
Insoluble matter=<0.01 % (in dilute H2SO4)
Alkalinitypasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only