Thermo Scientific Chemicals

Zinc oxide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 422745000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc oxide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 422745000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 422745000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-13-2
IUPAC Name
oxozinc
Molecular Formula
OZn
InChI Key
XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES
O=[Zn]
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Titration Complexometric
>=99.0 %
Insoluble matter
=<0.01 % (in dilute H2SO4)
Alkalinity
passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text