Copper(II) sulfate, 98%, pure, anhydrous
Copper(II) sulfate, 98%, pure, anhydrous
Copper(II) sulfate, 98%, pure, anhydrous
Copper(II) sulfate, 98%, pure, anhydrous
Thermo Scientific Chemicals

Copper(II) sulfate, 98%, pure, anhydrous

CAS: 7758-98-7 | CuO4S | 159.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 g
2.5 kg
Packaging:
Glass bottle
Catalog number 422871000
Price (USD)
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
63.20
Save 11.55 (18%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
63.20
Save 11.55 (18%)
Each
Add to cart
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Iodometric>=97.5 %
Chloride (Cl)=<50 ppm
Appearance (Color)Light grey
Iron (Fe)=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link