1x1 image pixel for data collection
Not web orderable
The product 422980100: Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced is not orderable at the moment. Please contact our Technical Support for possible alternatives.
Explore other products in this category
Palladium (Pd)
<p>An element belonging to the platinum group of metals, palladium (Kr4d<sup>10<sup>) (CAS 7440-05-3) is steel-white in color, except in powder form when it appears black. Palladium resists tarnishing in air and, if annealed, is soft and ductile.&nbsp;Palladium has the lowest density and melting point of the palladium metals and is the most reactive. At room temperature, palladium absorbs up to 900 times its own volume of hydrogen.<p> <p><a href="searchbrowsecategoryusen80014055">Transition Metal Salts <a>&gt; <a href="searchbrowsecategoryusen80014042">Metal Alloys&gt;<a><p> <p><strong>Phase at STP: <strong>Solid<p> <p><strong>Common Oxidation States: <strong>+2,4<p> <p><strong>Number of Valence Electrons:<strong> 10<p> <p><strong>Electronegativity (Pauling): <strong>2.2<p> <p><strong>First Ionization Energy: <strong>192 kcalmole<p> <p><strong>Covalent Radius: <strong> 1.28 Angstroms<p> <p><strong>Melting Point: <strong>1554.9&deg;C<p> <p><strong>Boiling Point: <strong>3167&deg;C<p> <p><strong>Density: <strong>12.02 gcm<sup>3<sup><p> <p><strong>Crystal Structure: <strong>Cubic, face-centered<p>