Thermo Scientific Chemicals

Potassium nitrite, 96+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423065000
500 g, Each
Thermo Scientific Chemicals

Potassium nitrite, 96+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423065000
500 g, Each
Quantity
Catalog number: 423065000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-09-0
IUPAC Name
potassium nitrite
Molecular Formula
KNO2
InChI Key
BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES
[K+].[O-]N=O
Titration with KMnO4
>=96.0 %
Magnesium (Mg)
=<20 ppm
Appearance (Color)
White to light yellow
Heavy metals (as Pb)
=<10 ppm
Sulfate (SO4)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text