Thermo Scientific Chemicals

Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423150010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423150010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423150010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6153-56-6
IUPAC Name
oxalic acid dihydrate
Molecular Formula
C2H6O6
InChI Key
GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES
O.O.OC(=O)C(O)=O
Heavy metals (ICP-OES)
=<5 ppm
Residue after ignition
=<0.01 %
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with KMnO4
99.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text