Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Thermo Scientific Chemicals

Oxalic acid dihydrate, 99.5+%, ACS reagent

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 423152500
Price (USD)
69.40
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
69.40
Each
Add to cart
Chemical Identifiers
CAS6153-56-6
IUPAC Nameoxalic acid dihydrate
Molecular FormulaC2H6O6
InChI KeyGEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILESO.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (ICP-OES)=<5 ppm
Residue after ignition=<0.01 %
Appearance (Color)White
Infrared spectrumConforms
Titration with KMnO499.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link