Thermo Scientific Chemicals

11-Bromoundecanoic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 423161000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

11-Bromoundecanoic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 423161000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2834-05-1
IUPAC Name
11-bromoundecanoic acid
Molecular Formula
C11H21BrO2
InChI Key
IUDGNRWYNOEIKF-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCCCCBr
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Color)
White to beige
Appearance (Form)
Crystalline chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text