Thermo Scientific Chemicals

Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 423175000
500 mL, EcoSafPak™ Glass Bottle, Each
Thermo Scientific Chemicals

Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 423175000
500 mL, EcoSafPak™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 423175000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Color scale
=<10 APHA
UV
at 255 nm A: =<1
UV
at 257 nm A: =<0.5
UV
at 263 nm A: =<0.1
UV
at 275 nm A: =<0.05

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text