Chemical Identifiers
IUPAC Nametris(1,1,1-trifluoro-4-hydroxy-4-(thiophen-2-yl)but-3-en-2-one) erbium hydrate
Molecular FormulaC24H17ErF9O7S3
InChI KeyLSMLVRQTTAIDBZ-UHFFFAOYSA-N
SMILESO.[Er].OC(=CC(=O)C(F)(F)F)C1=CC=CS1.OC(=CC(=O)C(F)(F)F)C1=CC=CS1.OC(=CC(=O)C(F)(F)F)C1=CC=CS1
Molecular Weight (g/mol)851.82
View more