Thermo Scientific Chemicals

1-Pentanesulfonic acid, sodium salt monohydrate, 98+%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 423200250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

1-Pentanesulfonic acid, sodium salt monohydrate, 98+%, HPLC grade, Thermo Scientific Chemicals

Catalog number: 423200250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 423200250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207605-40-1
IUPAC Name
pentane-1-sulfonate
Molecular Formula
C5H11O3S
InChI Key
RJQRCOMHVBLQIH-UHFFFAOYSA-M
SMILES
CCCCCS([O-])(=O)=O
UV
at 260 nm A: =<0.01
Appearance (Color)
Colorless to white
Appearance (Form)
Powder or crystals or flakes
Infrared spectrum
Conforms
Titration after Ion exchange
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text