Chemical Identifiers
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol)474.37
View more