Acetic acid, ammonium salt, 97+%, ACS reagent, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 97+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, ammonium salt, 97+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2 kg
Packaging:
Plastic bottle
Catalog number 423270250
Price (USD)/ Each
36.65
Online exclusive
42.60 
Save 5.95 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
42.60 
Save 5.95 (14%)
Add to cart
Acetic acid, ammonium salt, 97+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423270250
Price (USD)/ Each
36.65
Online exclusive
42.60 
Save 5.95 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
Molecular Weight (g/mol)77.08
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Chloride (Cl)=<5 ppm
Appearance (Color)White
Appearance (Form)Adhering crystals
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only