Thermo Scientific Chemicals

Ammonium fluoride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423295000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium fluoride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423295000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423295000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-01-8
IUPAC Name
ammonium hydrogen difluoride
Molecular Formula
F2H5N
InChI Key
KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES
[H+].[NH4+].[F-].[F-]
Appearance (Color)
White
Titration excess NaOH
>=98.0 %
Heavy metals (as Pb)
=<5 ppm
Insoluble matter
=<0.005 %
Residue after ignition
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text